Learn More
Medchemexpress LLC Roflumilast N-oxide | 292135-78-5 | 99.5% | 419.21 | C17H14Cl2F2N2O4 | 10 MG

Supplier: Medchemexpress LLC HY10063910MG
Roflumilast N-oxide is the N-oxide metabolite and analytical standard of roflumilast, acting as a phosphodiesterase 4 (PDE4) inhibitor for research and analytical applications, including biochemical assays and pharmacology studies.
- High purity (99.52%).
- Molecular formula C17H14Cl2F2N2O4 and molecular weight 419.21.
- Available as powder and as a 10 mM solution in DMSO.
- Suitable for use as an analytical standard and in in vitro assays.
- Recommended storage: powder at -20°C; in solvent at -80°C for long term.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.