Learn More
Medchemexpress LLC Roflumilast N-oxide | 292135-78-5 | MFCD04112984 | 99.2% | 419.21 g/mol | C17H14Cl2F2N2O4 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY100639R10MG
Encompass_Preferred
Roflumilast N-oxide (Standard) is an analytical reference standard of Roflumilast N-oxide intended for research and analytical applications. Supplied as a white solid with a certificate of analysis, the material is characterized for purity, identity, and stability and should be stored at -20°C protected from light and moisture.
Catalog No. 50-002-14466