Learn More
Medchemexpress LLC Roflumilast n-oxide | 292135-78-5 | MFCD04112984 | 99.5% | 419.21 | C17H14Cl2F2N2O4 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10063950MG
Encompass_Preferred
Roflumilast n-oxide is the N-oxide metabolite of roflumilast and a selective phosphodiesterase 4 (PDE4) inhibitor provided for research and analytical applications. It is available as solid quantities or as ready-to-use 10 mM DMSO solutions, and is intended for use in biochemical assays, compound characterization, and pharmacological studies.
  • Active PDE4 inhibitor and primary metabolite of roflumilast.
  • Supplied as solid quantities and 10 mM DMSO solutions for flexibility.
  • High purity (99.52%) suitable for analytical and research use.
  • Molecular formula C17H14Cl2F2N2O4 and molecular weight 419.21.
  • Useful as an analytical standard and for studies of inflammatory signaling.
  • SDS and certificate of analysis available to support safe handling and reproducibility.
Catalog No. 50-002-14465