Learn More
Medchemexpress LLC Roflumilast N-oxide | 292135-78-5 | MFCD04112984 | 99.5% | 419.21 g/mol | C17H14Cl2F2N2O4 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY100639R1MG
Encompass_Preferred
Roflumilast N-oxide is the N-oxide metabolite of roflumilast and acts as a phosphodiesterase type 4 (PDE4) inhibitor. Supplied as an analytical standard, this compound is intended for research and analytical applications and is not for human or clinical use. The product is typically supplied with high purity and characterized identity for laboratory use.
  • Analytical standard suitable for method development and reference.
  • High purity for reliable quantitative analysis.
  • Well-characterized chemical identity (CAS 292135-78-5).
  • Available in small solid quantities and solution formats for convenience.
  • Intended for research and analytical use only.
Catalog No. 50-002-14281