Learn More
Medchemexpress LLC Roflumilast N-oxide | 292135-78-5 | MFCD04112984 | 99.5% | 419.21 g/mol | C17H14Cl2F2N2O4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY100639R5MG
Encompass_Preferred
Roflumilast N-oxide is the analytical standard and active metabolite of roflumilast, a phosphodiesterase-4 (PDE4) inhibitor. Supplied for research and analytical applications, it is used as a reference standard in method development and for quantitative LC-MS and HPLC analyses.
  • Analytical standard for method development and quantitation.
  • High purity (99.52%) suitable for reference use.
  • Available as solid and as a 10 mM solution in DMSO for flexibility.
  • Includes downloadable COA, SDS, HNMR, LCMS, and handling instructions.
  • Suitable for LC-MS and HPLC assay validation and calibration.
Catalog No. 50-002-14187