Learn More
Selleck Chemical LLC Roflumilast N-oxide S0123-25MG
SDP
Supplier:  Selleck Chemical LLC S012325MG
Encompass_Preferred
Roflumilast N-oxide is an inhibitor of PDE type 4
Catalog No. 50-000-40294