Learn More
Medchemexpress LLC Roflupram 10mM 1mL | 1093412-18-0 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY11538310MM 1ML
Encompass_Preferred
Roflupram is a selective, orally active, brain-penetrant phosphodiesterase 4 (PDE4) inhibitor used in research to probe PDE4-mediated signaling and neuropharmacology. It is provided as a ready-to-use solution for laboratory applications and is intended for research use only.
  • Selective PDE4 inhibitor with reported IC50 ≈ 26.2 nM against human PDE4 catalytic domains.
  • Brain-penetrant and orally active in preclinical studies.
  • Supplied as 10 mM solution in DMSO, 1 ML volume for immediate use in assays.
  • High purity (reported ~99.45%) and well-characterized molecular properties (C16H20F2O4; MW 314.32 g/mol).
  • Soluble in DMSO at high concentration, suitable for in vitro dosing and formulation work.
Catalog No. 50-002-26715