Learn More
Medchemexpress LLC Roflupram 5mg | 1093412-18-0 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1153835MG
Encompass_Preferred
Roflupram is a selective, orally active, brain-penetrant phosphodiesterase 4 (PDE4) inhibitor used as a research compound for preclinical pharmacology and biochemical studies. Packaged as a 5 mg solid for laboratory use.
  • Selective PDE4 inhibition with reported IC50 26.2 nM
  • Brain-penetrant and orally active in preclinical models
  • Reported to reverse cognitive deficits and reduce pro-inflammatory factor production
  • High purity suitable for biochemical assays
  • Soluble in DMSO for solution preparations
Catalog No. 50-003-07806