Learn More
Medchemexpress LLC Roginolisib hemifumarate | 1621688-31-0 | 99.8% | C26H27FN4O5S.1/2C4H4O4 | 10MG
SDP
Supplier:  Medchemexpress LLC HY135827A10MG
Encompass_Preferred
Roginolisib hemifumarate is an orally active, selective PI3Kδ inhibitor used in preclinical and clinical research. It acts via a non-ATP-competitive mechanism with a reported IC50 around 145 nM and demonstrates high selectivity across a broad kinase panel. The compound is supplied as the hemifumarate salt and is intended for biochemical and cellular studies (research use only).
  • Selective PI3Kδ inhibition with reported IC50 ≈145 nM.
  • High selectivity versus a broad panel of kinases.
  • Orally active profile suitable for in vivo research.
  • Supplied as a hemifumarate salt for consistent formulation.
  • High purity appropriate for biochemical and cellular assays.
Catalog No. 50-002-58368