Learn More
Medchemexpress LLC HY-14751 5mg Medchemexpress, Rolapitant CAS:552292-08-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY147515MG
Encompass_Preferred
Medchemexpress, HY-14751 5mg Rolapitant CAS:552292-08-7 Rolapitant (SCH619734) is a potent, selective and orally active neurokinin NK1 receptor antagonist with a Ki of 0.66 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1635