Learn More
TARGETMOL CHEMICALS INC ROLAPITANT HYDROCHLORIDE 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T37245MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Rolapitant (SCH619734) hydrochloride is a potent selective long-acting and orally active neurokinin 1 (NK1) receptor antagonist with a K i of 0.66 nM. Rolapitant hydrochloride (Rolapitant HCl) does not interact with CYP3A4. Rolapitant hydrochloride shows potent centrally-mediated anti-emetic activity in both acute and delayed ferret emesis models [1] [2]. purity: 99%
Catalog No. 50-258-0143