Learn More
Selleck Chemical LLC Rolipram ZK-62711-10mg
SDP
Supplier:  Selleck Chemical LLC S143010MG
Encompass_Preferred
The PDE4 selective inhibitor, Rolipram (ZK-62711, SB 95952), inhibited immunopurified PDE4B and PDE4D activities similarly, with IC50 values of approx. 130 nM and 240 nM respectively; an anti-inflammatory agent.
Catalog No. 50-238-9975