Learn More
Essendant ROLLATM3.125X19604CNWE
Supplier:  Essendant ICX90930002
Encompass_Preferred
The paper rolls are guaranteed to work efficiently with your machine printhead to provide crisp clear black images The reliability and performance of a thermal printer is directly related to the quality of the supplies including the paper used by the customer The IBM Testing Process is an Industry Standard test for the thermal papers used in the marketplace To be IBM approved the paper must pass a 100km of paper printhead test without abrasion or build-up with no cutting failures IBM testing only confirms that the use of approved papers will not negatively affect the operating life of an IBM thermal printer Manufactured to OEM specifications ATM Rolls-Thermal receipt with coated side in Sheet/Roll Size- 3 13x1960 ft Paper/Media Color(s)- White Color Family- White Machine Compatibility- ATMs
Catalog No. 50-000-33709