Learn More
Essendant ROLLTHML2 5/16X400WH
Supplier:  Essendant ICX90782978
Encompass_Preferred
The paper rolls are guaranteed to work efficiently with your machine printhead to provide crisp clear black images The reliability and performance of a thermal printer is directly related to the quality of the supplies including the paper used by the customer The IBM Testing Process is an Industry Standard test for the thermal papers used in the marketplace To be IBM approved the paper must pass a 100km of paper printhead test without abrasion or build-up with no cutting failures IBM testing only confirms that the use of approved papers will not negatively affect the operating life of an IBM thermal printer Manufactured to OEM specifications Sheet/Roll Size- 2 31x400 ft Paper/Media Color(s)- White Color Family- White Machine Compatibility- POS Machines
Catalog No. 50-000-33705