Learn More
Medchemexpress LLC Roluperidone | 359625-79-9 | 99.9% | 366.43 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1946950MG
Encompass_Preferred
Roluperidone is a novel cyclic amide derivative exhibiting high equipotent affinities for 5-HT2A and sigma-2 receptors. It also shows binding affinity for α1-adrenergic receptors, with low or no affinity for muscarinic, cholinergic, and histaminergic receptors. Sigma-2 receptors are likely involved in modulating dopamine and glutamatergic pathways, as well as calcium neuronal modulation.
  • High equipotent affinities for 5-HT2A and sigma-2 receptors.
  • Exhibits binding affinity for α1-adrenergic receptors.
  • Low or no affinity for muscarinic, cholinergic, and histaminergic receptors.
  • Involved in modulating dopamine and glutamatergic pathways.
  • Plays a role in calcium neuronal modulation.
Catalog No. 50-003-85081