Learn More
Medchemexpress LLC HY-15149 10mg , Romidepsin FK 228;FR 901228;NSC 630176 CAS:128517-07-7 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY1514910MG
Encompass_Preferred
Medchemexpress, HY-15149 10mg Romidepsin FK 228;FR 901228;NSC 630176 CAS:128517-07-7 Formula:C24H36N4O6S2 IC50: 36 nM (HDAC1), 47 nM (HDAC2) Purity:98% Romidepsin is a potent  HDAC1  and  HDAC2  inhibitor with  IC 50 values of 36 and 47 nM, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-4037
 Restricted Item