Learn More
Apexbio Technology LLC Romidepsin (FK228, depsipeptide), 5mg. Cas: 128517-07-7 MFCD: MFCD18433404. HDAC1/HDAC2 inhibitor, potent and selective
SDP
Supplier:  Apexbio Technology LLC A81735
Encompass_Preferred
Romidepsin, also known as FK228 or depsipeptide, is potent and selective inhibitor of histone deacetylases (HDACs) which are associated with the regulation of re-expression of silenced tumor suppressor genes. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5068