Learn More
Cayman Chemical RomIfIdInhydrochlorIde 10mg
SDP
Supplier:  Cayman Chemical 3669310 MG
Encompass_Preferred
An α2-AR agonist; epidural administration induces antinociception and sedation in donkeys at 50 UG/kg; induces bradycardia in horses from 0.04-0.12 mg/kg
Catalog No. 50-291-3853