Learn More
Medchemexpress LLC Roniciclib | 1223498-69-8 | 98.0% | 430.44 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY13914
Encompass_Preferred
Roniciclib is an orally bioavailable pan-cyclin dependent kinase (CDK) inhibitor. It has IC50s of 5-25 nM for CDK1, CDK2, CDK3, CDK4, CDK7, and CDK9.
  • Inhibits the kinase activity of cell-cycle CDKs (CDK1/cyclin B, CDK2/cyclin E, and CDK4/cyclin D)
  • Inhibits transcriptional CDKs (CDK9/cyclin T1 and CDK7/cyclin H/MAT1)
  • Potently inhibits the proliferation of various human and murine tumor cell lines
  • Strongly inhibits tumor growth in a dose-dependent manner
  • Low blood clearance rates in mouse, rat, and dog
Catalog No. 50-003-48124