Learn More
Apexbio Technology LLC Ronidazole 7681-76-7 5g
SDP
Supplier:  Apexbio Technology LLC B20225000
Encompass_Preferred
Ronidazole is a nitroimidazole compound exhibiting both antiprotozoal and antibacterial activities Its mechanism of action involves the generation of metabolites that interact with microbial DNA resulting in DNA damage and inhibition of nucleic acid synthesis Ronidazole is commonly employed in veterinary research settings to investigate therapeutic interventions against protozoan parasites such as Tritrichomonas foetus infections in feline models Histomonas meleagridis-mediated histomoniasis and Brachyspira hyodysenteriae-related swine dysentery It can be administered orally to experimental animal subjects for clarifying pathological mechanisms and therapeutic pathways in parasitic disease models Currently detailed IC50 data for Ronidazole remain underreported in literature
Catalog No. 50-000-62445