Learn More
Cayman Chemical RopIdoxurIdIn 10mg
SDP
Supplier:  Cayman Chemical 4122010 MG
Encompass_Preferred
A radiosensitizer and a prodrug form of idoxuridine; inhibits viral plaque formation in HSV-1- or HSV-2-HeLa infected cells at 100 µM; enhances radiation-induced decreases in tumor growth without reducing body weight in an HT-29 colon cancer mouse xenograft model at 1,000 mg/kg per day for six days; increases animal survival in a mouse model of lethal HSV-2 infection at 100 mg/kg every five days
Catalog No. 50-294-4536