Learn More
Selleck Chemical LLC Ropinirole HCl S3189-25mg
SDP
Supplier:  Selleck Chemical LLC S318925MG
Encompass_Preferred
Ropinirole (SKF-101468A) a selective dopamine D2 receptors agonist with Ki of 29 nM
Catalog No. 50-310-9958