Learn More
TARGETMOL CHEMICALS INC Ropinirole hydrochloride 200MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T2592200MG
Encompass_Preferred
Also available in 1 mL, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Ropinirole hydrochloride (SKF-101468A) is a selective dopamine D2 receptors agonist (Ki 29 nM). Ropinirole hydrochloride (SKF-101468A) is the hydrochloride salt form of ropinirole, a non-ergot dopamine agonist with antiparkinsonian property. Purity 99.98%
Catalog No. 50-287-7505