Learn More
Medchemexpress LLC Ropitoin (TR 2985) | 56079-81-3 | 483.60 | 20 MG

Supplier: Medchemexpress LLC HYU0027420MG
Ropitoin (TR 2985) is a novel antiarrhythmic agent.
- Purity: 98.2%
- Molecular weight: 483.60
- Chemical formula: C₃₀H₃₃N₃O₃
- CAS number: 56079-81-3
- Appearance: Solid
- Color: Off-white to light yellow
- Shipping: Room temperature in continental US; may vary elsewhere.
- Storage (powder): -20°C for 3 years, 4°C for 2 years.
- Storage (in solvent): -80°C for 6 months, -20°C for 1 month.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.