Learn More
Medchemexpress LLC Ropivacaine-d7 | 1392208-04-6 | 99.6% | 281.44 | C17H19D7N2O | 10 MG
SDP
Supplier:  Medchemexpress LLC HYB0563S10MG
Encompass_Preferred
(Rac)-Ropivacaine-d7 is the deuterium-labeled racemic form of ropivacaine intended for use as a stable isotope tracer or internal standard in quantitative analytical workflows. It is suitable for NMR, GC-MS, and LC-MS method development, quantitation, and metabolic studies, and is provided as a high-purity neat material with supporting documentation.
  • Deuterium-labeled (d7) racemic ropivacaine for mass-shifted analysis.
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS quantitation.
  • High chemical purity: 99.6%.
  • Molecular formula C17H19D7N2O; molecular weight 281.44.
  • Available in small quantities for analytical use, for example 10 mg.
  • Supplied with data sheet, certificate of analysis, and safety data sheet.
Catalog No. 50-002-99031