Learn More
Medchemexpress LLC Ropivacaine-d7 | 684647-62-9 | 99.7% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB0563S15MG
Encompass_Preferred
Ropivacaine-d7 is deuterium labeled Ropivacaine. Ropivacaine is a potent sodium channel blocker, which blocks impulse conduction by reversibly inhibiting sodium ion influx in nerve fibers. It is also an inhibitor of K2P (two-pore domain potassium channel) TREK-1 with an IC50 of 402.7 μM in COS-7 cell's membrane. Ropivacaine is used for the research of neuropathic pain management. It can be used as a tracer or an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium labeled Ropivacaine
  • Potent sodium channel blocker
  • Inhibits impulse conduction by reversibly inhibiting sodium ion influx
  • Inhibitor of K2P (two-pore domain potassium channel) TREK-1
  • Used for research in neuropathic pain management
  • Can be used as a tracer or internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Related to isotope-labeled compounds
  • Useful for therapeutic drug monitoring
Catalog No. 50-004-08698