Learn More
Apexbio Technology LLC Ropivacaine HCl, 98717-15-8, MFCD02102164, 50 mg
SDP
Supplier:  Apexbio Technology LLC B202350
Encompass_Preferred
MF: C17H26N2OHCl, MW: 310.86, Solubility: 10.1mg/mL in H2O. Ropivacaine HCl is an anaesthetic agent and blocks impulse conduction in nerve fibres through inhibiting sodium ion influx reversibly.
Catalog No. 50-115-1531