Learn More
Selleck Chemical LLC Ropsacitinib S9676-100MG
SDP
Supplier:  Selleck Chemical LLC S9676100MG
Encompass_Preferred
Ropsacitinib (PF-06826647 Tyk2-IN-8 compound 10) is a selective and orally administered inhibitor of tyrosine kinase 2 (TYK2) with IC50 of 17 nM for binding to TYK2 catalytically active JH1 domain PF-06826647 (Tyk2-IN-8 compound 10) also inhibits JAK1 and JAK2 with IC50 of 383 nM and 74 nM respectively PF-06826647 (Tyk2-IN-8 compound 10) is used in the treatment of psoriasis (PSO)
Catalog No. 50-000-24419