Learn More
Medchemexpress LLC Roquefortine C | 58735-64-1 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN67481MG
Encompass_Preferred
Roquefortine C is a mycotoxin isolated from Penicillium species. It acts as an agonist of P-gp and an inhibitor of P450 3A and P450 1A. This compound can inhibit Gram-positive bacteria, exhibits neurotoxicity, and demonstrates antitumor activity.
  • Solid appearance
  • White to off-white color
  • Classified as a cyclopeptide
  • Isolated from microorganisms, specifically Penicillium roqueforti
Catalog No. 50-004-20225