Learn More
Medchemexpress LLC ROS kinases-IN-1 | 370096-57-4 | 98.3% | 328.37 | 1 ML
Supplier: Medchemexpress LLC HY14822810MM/1ML
ROS kinases-IN-1 is a ROS tyrosine kinase inhibitor with an IC50 value of 1.22 μM, demonstrating anti-tumor activity by inhibiting the growth of cervical cancer and glioblastoma cells.
- Acts as a ROS tyrosine kinase inhibitor
- Has an IC50 value of 1.22 μM for ROS kinase
- Shows anti-tumor activity
- Inhibits the growth of cervical cancer and glioblastoma cells
- Effective at concentrations of 1-100 μM over one day
- Exhibits EC50s for HeLa, U138, U87, LN18, and U118 cells