Learn More
Medchemexpress LLC Rosarin | 84954-93-8 | 428.43 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN05061MG
Encompass_Preferred
Rosarin is a cinnamyl alcohol glycoside isolated from Rhodiola rosea. It exhibits anti-inflammatory and neuroprotective effects, suppressing the expression of proinflammatory factors such as iNOS, IL-1β, and TNF-α in the kidney and prefrontal cortex of the brain in mice.
  • Isolated from Rhodiola rosea.
  • Exhibits anti-inflammatory effects.
  • Exhibits neuroprotective effects.
  • Suppresses expression of proinflammatory factors such as iNOS, IL-1β, and TNF-α.
  • Pretreatments with Rosarin can suppress iNOS and cytokine expression in murine BV2 cells.
Catalog No. 50-003-90207