Learn More
Medchemexpress LLC Rosarin | 84954-93-8 | 428.43 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN05065MG
Encompass_Preferred
Rosarin is a cinnamyl alcohol glycoside isolated from Rhodiola rosea. It exhibits anti-inflammatory and neuroprotective effects, suppressing the expression of pro-inflammatory factors such as iNOS, IL-1β, and TNF-α in the kidney and prefrontal cortex of the brain in mice.
  • Isolated from Rhodiola rosea.
  • Possesses anti-inflammatory effects.
  • Exhibits neuroprotective effects.
  • Suppresses the expression of pro-inflammatory factors iNOS, IL-1β, and TNF-α.
Catalog No. 50-003-88718