Learn More
Selleck Chemical LLC Roscovitine S1153-10mM/1mL
SDP
Supplier:  Selleck Chemical LLC S115310MM/1ML
Encompass_Preferred
Roscovitine is a potent and selective CDK inhibitor for Cdc2 CDK2 and CDK5 with IC50 of 0 65 M 0 7 M and 0 16 M in cell-free assays It shows little effect on CDK4/6 Phase 2
Catalog No. 50-310-8534