Learn More
Medchemexpress LLC HY-D0214 50mg , Rose Bengal (sodium) CAS:632-69-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYD021450MG
Encompass_Preferred
HY-D0214 50mg Rose Bengal (sodium) CAS: 632-69-9 Rose Bengal sodium, a synthetic fluorescein derivative, and is a crimson-coloured dye with the principal component being 4,5,6,7-tetrachloro-2,4,5,7-tetraiodo fluorescein. Rose Bengal sodium has been widely used as an ophthalmic diagnostic agents, and can detect desiccated or damaged cells on the ocular surface. Rose Bengal sodium exhibits antiviral activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1935