Learn More
STEMCELL Technologies RosetteSep™ Human B Cell Enrichment Cocktail, For processing: 200 mL whole blood processed
SDP
Supplier:  STEMCELL Technologies 15064
Encompass
The RosetteSep™ Human B Cell Enrichment Cocktail is designed to isolate B cells from whole blood by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-B cells and glycophorin A on red blood cells (RBCs). When centrifuged over a buoyant density medium such as RosetteSep™ DM-L (Catalog #15705) or Lymphoprep™ (Catalog #07801), the unwanted cells pellet along with the RBCs. The purified B cells are present as a highly enriched population at the interface between the plasma and the buoyant density medium.
Catalog No. NC0178711