Learn More
Selleck Chemical LLC Rosiglitazone maleate 25mg 155141-29-0 BRL 49653
SDP
Supplier:  Selleck Chemical LLC S250525MG
Encompass_Preferred
Rosiglitazone maleate (BRL 49653), a member of the thiazolidinedione class of antihyperglycaemic agents, is a high-affinity selective agonist of the peroxisome proliferator-activated receptor-? (PPAR-?) with IC50 of 42 nM. Rosiglitazone maleate also modulates TRP channels and induces autophagy. Rosiglitazone prevents ferroptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-607-6