Learn More
Medchemexpress LLC Rosmanol | 80225-53-2 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN501550MG
Encompass_Preferred
Rosmanol can inhibit the oxidation of low-density lipoprotein (LPL) and significantly inhibit lipopolysaccharide-induced iNOS and COX-2 expression, demonstrating an anti-inflammatory effect.
  • Inhibits oxidation of low-density lipoprotein (LPL)
  • Significantly inhibits lipopolysaccharide-induced iNOS expression
  • Significantly inhibits lipopolysaccharide-induced COX-2 expression
  • Demonstrates anti-inflammatory effects
Catalog No. 50-004-19780