Learn More
Cayman Chemical RostafuroxIn 10mg
SDP
Supplier:  Cayman Chemical 2128210 MG
Encompass_Preferred
Prevents binding of endogenous ouabain to the Na+/K+ ATPase (IC50 = 1.7 µM)
Catalog No. 50-298-5048