Learn More
Medchemexpress LLC Rostafuroxin | 156722-18-8 | ≥98.0% | 100 MG

Supplier: Medchemexpress LLC HY12283100MG
Rostafuroxin is a small-molecule Na+/K+-ATPase (ATP1A1) antagonist supplied as an analytical research standard with reported antihypertensive and anti-RSV activity.
- Analytical standard for Na+/K+-ATPase (ATP1A1) research.
- Molecular formula C23H34O4; molecular weight 374.51 g/mol.
- Reported antihypertensive and anti-RSV activity.
- Typical purity ≥98% (HPLC).
- Commonly supplied as a 100 mg laboratory standard.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.