Learn More
Apexbio Technology LLC Rosuvastatin Calcium 147098-20-2 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC A85185.1
Encompass_Preferred
Rosuvastatin Calcium (147098-20-2) is a small-molecule inhibitor targeting 3-hydroxy-3-methylglutaryl-coenzyme A (HMG-CoA) reductase It is designed to suppress this enzyme s activity thereby regulating cholesterol biosynthesis and lipid metabolism Rosuvastatin Calcium exerts its biological activity primarily through competitive inhibition of HMG-CoA reductase In biochemical assays Rosuvastatin Calcium demonstrates potent inhibition with an IC50 value of approximately 5 nM for human liver HMG-CoA reductase Based on these pharmacological properties Rosuvastatin Calcium holds research potential in cholesterol-related pathway studies regulation of lipid metabolism and investigation of cardiovascular conditions
Catalog No. 50-000-95269