Learn More
Medchemexpress LLC Rosuvastatin (Calcium) | 147098-20-2 | 99.8% | 500.57 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1750410MM/1ML
Encompass_Preferred
Rosuvastatin Calcium is a competitive HMG-CoA reductase (HMGCR) inhibitor that potently blocks hERG current. It effectively lowers low-density lipoprotein (LDL) cholesterol, triglycerides, and C-reactive protein levels.
  • Competitive HMG-CoA reductase (HMGCR) inhibitor.
  • Potently blocks hERG current.
  • Reduces expression of mature hERG and interaction with heat shock protein 70 (Hsp70).
  • Effectively lowers LDL cholesterol, triglycerides, and C-reactive protein levels.
Catalog No. 50-003-86780