Learn More
Medchemexpress LLC Rosuvastatin (calcium) | 147098-20-2 | 99.8% | 500.57 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY17504R5MG
Encompass_Preferred
Rosuvastatin Calcium is a competitive HMG-CoA reductase (HMGCR) inhibitor, with an IC50 of 11 nM. It potently blocks hERG current with an IC50 of 195 nM. Rosuvastatin Calcium reduces the expression of the mature hERG and the interaction of heat shock protein 70 (Hsp70) with the hERG protein. This compound effectively lowers low-density lipoprotein (LDL) cholesterol, triglycerides, and C-reactive protein levels.
  • Competitive HMG-CoA reductase (HMGCR) inhibitor with an IC50 of 11 nM.
  • Potently blocks hERG current with an IC50 of 195 nM.
  • Reduces the expression of mature hERG and its interaction with Hsp70.
  • Effectively lowers low-density lipoprotein (LDL) cholesterol.
  • Effectively lowers triglycerides.
  • Effectively lowers C-reactive protein levels.
Catalog No. 50-003-87193