Learn More
Medchemexpress LLC Rosuvastatin Calcium | 147098-20-2 | 99.8% | 520.61 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY17504R50MG
Encompass_Preferred
Rosuvastatin Calcium is a competitive HMG-CoA reductase (HMGCR) inhibitor, with an IC50 of 11 nM. Rosuvastatin Calcium potently blocks hERG current with an IC50 of 195 nM. It reduces the expression of the mature hERG and the interaction of heat shock protein 70 (Hsp70) with the hERG protein. Rosuvastatin Calcium effectively lowers low-density lipoprotein (LDL) cholesterol, triglycerides, and C-reactive protein levels.
  • Competitive HMG-CoA reductase (HMGCR) inhibitor with an IC50 of 11 nM
  • Potently blocks hERG current with an IC50 of 195 nM
  • Reduces the expression of the mature hERG and the interaction of heat shock protein 70 (Hsp70) with the hERG protein
  • Effectively lowers low-density lipoprotein (LDL) cholesterol, triglycerides, and C-reactive protein levels
Catalog No. 50-003-87041