Learn More
Medchemexpress LLC Rosuvastatin calcium | 147098-20-2 | MFCD18783208 | 99.9% | 520.61 g·mol⁻¹ | C44H54CaF2N6O12S2 | 10 MG

Supplier: Medchemexpress LLC HY17504R10MG
Rosuvastatin calcium (standard) is an analytical reference material supplied as the calcium salt for research and analytical use. It is a characterized, high-purity material intended for assay, method development, calibration, and quality-control workflows in laboratory environments.
- High purity: 99.9%.
- Molecular weight: 520.61 g·mol⁻¹.
- Chemical formula: C44H54CaF2N6O12S2.
- CAS number: 147098-20-2.
- Intended for analytical, assay, and QC applications.
- Available in multiple pack sizes, including 10 mg.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.