Learn More
Selleck Chemical LLC Rosuvastatin calcium S2169-10mM/1mL
SDP
Supplier:  Selleck Chemical LLC S216910MM/1ML
Encompass_Preferred
Rosuvastatin calcium is a competitive inhibitor of HMG-CoA reductase with IC50 of 11 nM in a cell-free assay
Catalog No. 50-311-1761