Learn More
TARGETMOL CHEMICALS INC Rosuvastatin calcium 500MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T1510500MG
Encompass_Preferred
Also available in 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Rosuvastatin calcium (ZD4522) , a selective and competitive inhibitor of hepatic hydroxymethyl-glutaryl coenzyme A (HMG-CoA) reductase, has antilipidemic activity. Purity 99.68%
Catalog No. 50-287-8437