Learn More
Medchemexpress LLC Rosuvastatin (Calcium) (Standard) | 147098-20-2 | 99.8% | 520.61 | 25MG
SDP
Supplier:  Medchemexpress LLC 25MGHY17504R
Encompass_Preferred
Rosuvastatin Calcium is a competitive HMG-CoA reductase (HMGCR) inhibitor, with an IC50 of 11 nM. It potently blocks hERG current with an IC50 of 195 nM. This compound reduces the expression of the mature hERG and the interaction of heat shock protein 70 (Hsp70) with the hERG protein. Rosuvastatin Calcium effectively lowers low-density lipoprotein (LDL) cholesterol, triglycerides, and C-reactive protein levels. It is for research use only.
  • Competitive HMG-CoA reductase (HMGCR) inhibitor
  • Potently blocks hERG current
  • Reduces expression of mature hERG and interaction with Hsp70
  • Effectively lowers LDL cholesterol, triglycerides, and C-reactive protein levels
Catalog No. 50-003-84389