Learn More
Medchemexpress LLC Rosuvastatin lactone | 503610-43-3 | 97.0% | 463.52 | 25 MG

Supplier: Medchemexpress LLC 25MGHY138166
Rosuvastatin lactone is a metabolite of Rosuvastatin and an HMG-CoA inhibitor. It is intended for research use only.
- Metabolite of Rosuvastatin
- HMG-CoA inhibitor
- For research use only
- Powder storage: -20°C for 3 years, 4°C for 2 years
- In solvent storage: -80°C for 6 months, -20°C for 1 month
- Soluble in DMSO (125 mg/mL)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.