Learn More
Medchemexpress LLC Rosuvastatin lactone | 503610-43-3 | 98.6% | 463.52 | 50 MG

Supplier: Medchemexpress LLC HY13816650MG
Rosuvastatin lactone is a metabolite of Rosuvastatin, an HMG-CoA inhibitor. It is intended for research use only and is not sold to patients. This product appears as a white to off-white solid.
- Purity: 98.6%
- Molecular weight: 463.52
- Chemical formula: C22H26FN3O5S
- Shipping: Room temperature in continental US
- Storage: Powder at -20°C for 3 years or 4°C for 2 years
- Storage: In solvent at -80°C for 6 months or -20°C for 1 month
- Solubility: 125 mg/mL in DMSO
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.