Learn More
Medchemexpress LLC Rosuvastatin sodium | 147098-18-8 | 99.9% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY17504B100MG
Encompass_Preferred
Rosuvastatin Sodium is a competitive HMG-CoA reductase (HMGCR) inhibitor. It effectively lowers low-density lipoprotein (LDL) cholesterol, triglycerides, and C-reactive protein levels. It also potently blocks hERG current.
  • Potently blocks hERG current
  • Reduces expression of mature hERG and interaction of heat shock protein 70 (Hsp70)
  • Effectively lowers low-density lipoprotein (LDL) cholesterol
  • Lowers triglycerides
  • Lowers C-reactive protein levels
Catalog No. 50-003-87028